siRNA Details
Gene Symbol: TMEM150B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085488.2 | NP_001078957.1 | 6 | 441 | ||
NM_001282011.1 | NP_001268940.1 | 6 | 490 | |||
NR_104066.1 | 5 | 422 | ||||
NR_104071.1 | 5 | 373 | ||||
XM_005258812.3 | XP_005258869.1 | 8 | 770 | |||
XM_011526850.2 | XP_011525152.1 | 7 | 649 | |||
XM_011526851.2 | XP_011525153.1 | 7 | 647 | |||
XM_011526852.2 | XP_011525154.1 | 7 | 647 | |||
XM_011526853.2 | XP_011525155.1 | 6 | 561 | |||
XM_011526854.2 | XP_011525156.1 | 5 | 726 | |||
XM_011526855.2 | XP_011525157.1 | 7 | 599 | |||
XM_011526856.2 | XP_011525158.1 | 5 | 678 | |||
XM_011526857.2 | XP_011525159.1 | 4 | 341 | |||
XM_017026667.1 | XP_016882156.1 | 7 | 661 | |||
XM_017026668.1 | XP_016882157.1 | 4 | 364 | |||
GenBank mRNA | AU311823.1 | - | 6 | 441 | ||
BC171893.1 | - | 5 | 338 | |||
BC171901.1 | - | 5 | 387 | |||
BC171904.1 | - | 6 | 451 | 9054558 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
