siRNA Details
Gene Symbol: ZFAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003606.2 | 2, 3 | 650 | |||
GenBank mRNA | CB137861.1 | - | 2, 3 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
