siRNA Details
Gene Symbol: ZFAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003604.2 | 2 | 618 | |||
NR_003605.1 | 2 | 299 | ||||
NR_003606.2 | 2 | 618 | ||||
NR_036658.1 | 2 | 618 | ||||
NR_036659.1 | 2 | 176 | ||||
GenBank mRNA | AY339614.1 | - | 2 | 120 | ||
AY513722.1 | - | 2 | 618 | |||
AY513723.1 | - | 2 | 299 | |||
BC105282.1 | - | 2 | 158 | |||
BM745942.1 | - | 2 | 178 | |||
CB137861.1 | - | 2 | 154 | |||
CR936805.1 | - | 2 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
