siRNA Details
Gene Symbol: LINC01128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047519.1 | 6 | 4813 | |||
NR_047521.1 | 5 | 4660 | ||||
NR_047523.1 | 4 | 4564 | ||||
NR_047524.1 | 4 | 4476 | ||||
NR_047525.1 | 5 | 4527 | ||||
GenBank mRNA | AK096570.1 | - | 1 | 1473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
