siRNA Details
Gene Symbol: LINC01128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047519.1 | 6 | 5511 | |||
NR_047521.1 | 5 | 5358 | ||||
NR_047523.1 | 4 | 5262 | ||||
NR_047524.1 | 4 | 5174 | ||||
NR_047525.1 | 5 | 5225 | ||||
GenBank mRNA | AK091100.1 | - | 1 | 650 | ||
AK096570.1 | - | 1 | 2171 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
