siRNA Details
Gene Symbol: ZNF518B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053042.2 | NP_444270.2 | 3 | 1793 | ||
XM_005248193.2 | XP_005248250.1 | 2 | 1628 | |||
XM_017008784.1 | XP_016864273.1 | 2 | 4251 | |||
XM_017008785.1 | XP_016864274.1 | 3 | 2019 | |||
XM_017008786.1 | XP_016864275.1 | 2 | 1853 | |||
GenBank mRNA | AB051516.1 | - | 1 | 1145 | ||
AK122755.1 | - | 1 | 480 | |||
AL833306.1 | - | 1 | 1329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
