siRNA Details
Gene Symbol: LOC101929762 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125931.1 | 3 | 1842 | |||
GenBank mRNA | AK098126.1 | - | 4 | 2025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
