siRNA Details
Gene Symbol: ANKS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518357.2 | XP_011516659.1 | 1 | 2338 | ||
XM_017014446.1 | XP_016869935.1 | 1 | 2338 | |||
GenBank mRNA | BC012981.2 | - | 1 | 1353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
