siRNA Details
Gene Symbol: ZNF521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308225.1 | NP_001295154.1 | 2 | 331 | ||
NM_015461.2 | NP_056276.1 | 3 | 372 | |||
XM_011525909.2 | XP_011524211.1 | 3 | 372 | |||
XM_011525910.2 | XP_011524212.1 | 3 | 372 | |||
XM_011525911.1 | XP_011524213.1 | 4 | 408 | |||
XM_017025697.1 | XP_016881186.1 | 2 | 261 | |||
XM_017025698.1 | XP_016881187.1 | 4 | 335 | |||
GenBank mRNA | AJ518106.1 | - | 3 | 269 | ||
AK027354.1 | - | 3 | 274 | |||
AK293067.1 | - | 3 | 323 | |||
BC032869.2 | - | 2 | 230 | |||
BC113622.1 | - | 3 | 193 | 8322674 | ||
BC113648.1 | - | 3 | 193 | 8322700 | ||
BC143727.1 | - | 3 | 193 | |||
DA102816.1 | - | 3 | 372 |
