siRNA Details
Gene Symbol: AMIGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020703.2 | NP_065754.2 | 2 | 1334 | ||
XM_011541812.2 | XP_011540114.1 | 2 | 1340 | |||
GenBank mRNA | AB032989.1 | - | 1 | 786 | ||
AK295678.1 | - | 2 | 1313 | |||
AY237007.1 | - | 1 | 952 | |||
BC040879.1 | - | 2 | 1302 | 5725643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
