siRNA Details
Gene Symbol: SEPT7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024271.1 | 5 | 716 | |||
GenBank mRNA | AL832059.1 | - | 5 | 652 | ||
BC092503.1 | - | 5 | 661 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
