siRNA Details
Gene Symbol: SMG1P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308867.1 | - | 7 | 769 |
Gene Symbol: LOC107984138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023965.1 | XP_016879454.1 | 11 | 1410 | ||
XM_017023966.1 | XP_016879455.1 | 11 | 1410 | |||
XM_017023967.1 | XP_016879456.1 | 11 | 1410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
