siRNA Details
Gene Symbol: SLC38A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037984.2 | NP_001033073.1 | 16 | 3993 | ||
XM_005257019.1 | XP_005257076.1 | 17 | 4111 | |||
XM_011524288.1 | XP_011522590.1 | 17 | 4132 | |||
XM_011524289.1 | XP_011522591.1 | 16 | 4108 | |||
XM_011524290.1 | XP_011522592.1 | 15 | 3387 | |||
GenBank mRNA | AF086389.1 | - | 1 | 95 | ||
AF217993.1 | - | 1608 | ||||
BC011702.2 | - | 1 | 1381 | |||
BC014156.2 | - | 2 | 1651 | |||
BI194553.1 | - | 1 | 754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
