siRNA Details
Gene Symbol: PSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015310.3 | NP_056125.3 | 16 | 10421 | ||
NM_206909.2 | NP_996792.1 | 13 | 8825 | |||
XM_017013258.1 | XP_016868747.1 | 16 | 11730 | |||
XM_017013259.1 | XP_016868748.1 | 16 | 11708 | |||
XM_017013260.1 | XP_016868749.1 | 16 | 10481 | |||
XM_017013261.1 | XP_016868750.1 | 15 | 10695 | |||
XM_017013265.1 | XP_016868754.1 | 13 | 9231 | |||
GenBank mRNA | AK313826.1 | - | 1 | 682 | ||
AL353936.1 | - | 1 | 1606 | |||
BC011238.1 | - | 1 | 698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
