siRNA Details
Gene Symbol: LOC100130744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046285.1 | 1 | 2139 | |||
GenBank mRNA | AF289590.1 | - | 1 | 2159 | ||
AK129556.1 | - | 1 | 153 |
Gene Symbol: OTULIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011514151.2 | XP_011512453.1 | 8 | 7248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
