siRNA Details
Gene Symbol: XKR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052898.1 | NP_443130.1 | 2 | 1011 | ||
GenBank mRNA | AB067476.1 | - | 2 | 545 | ||
AY534241.1 | - | 2 | 1011 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
