siRNA Details
Gene Symbol: PRR14L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173566.2 | NP_775837.2 | 4 | 3523 | ||
XM_006724219.3 | XP_006724282.1 | 4 | 4083 | |||
XM_006724220.3 | XP_006724283.1 | 4 | 3485 | |||
XM_006724221.2 | XP_006724284.1 | 3 | 3039 | |||
XM_011530080.2 | XP_011528382.1 | 5 | 4291 | |||
XM_011530081.2 | XP_011528383.1 | 4 | 4167 | |||
XM_017028736.1 | XP_016884225.1 | 4 | 5243 | |||
XM_017028737.1 | XP_016884226.1 | 3 | 4681 | |||
GenBank mRNA | CT841517.1 | - | 3 | 3355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
