siRNA Details
Gene Symbol: MROH2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287395.1 | NP_001274324.1 | 42 | 5181 | ||
XM_011511075.2 | XP_011509377.1 | 42 | 5388 | |||
XM_011511076.2 | XP_011509378.1 | 42 | 5376 | |||
XM_011511077.2 | XP_011509379.1 | 42 | 5358 | |||
XM_011511078.2 | XP_011509380.1 | 42 | 5349 | |||
XM_011511079.2 | XP_011509381.1 | 42 | 5343 | |||
XM_011511080.2 | XP_011509382.1 | 42 | 6859 | |||
XM_011511081.2 | XP_011509383.1 | 41 | 5283 | |||
XM_011511082.2 | XP_011509384.1 | 41 | 5250 | |||
XM_011511084.2 | XP_011509386.1 | 41 | 5190 | |||
XM_011511085.2 | XP_011509387.1 | 36 | 4458 | |||
XM_011511086.2 | XP_011509388.1 | 33 | 4113 | |||
GenBank mRNA | BC042133.1 | - | 7 | 1008 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
