siRNA Details
Gene Symbol: FAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291285.1 | NP_001278214.1 | 1 | 4041 | ||
NM_001291303.1 | NP_001278232.1 | 1 | 4041 | |||
NM_024582.4 | NP_078858.4 | 1 | 4041 | |||
XM_011532236.2 | XP_011530538.1 | 2 | 5118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
