siRNA Details
Gene Symbol: PRR36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190467.1 | NP_001177396.1 | 6 | 4274 | ||
NR_104345.1 | 7 | 1617 | ||||
GenBank mRNA | AK025837.1 | - | 3 | 2010 | ||
AK096643.1 | - | 3 | 2286 | |||
AL442075.1 | - | 2 | 789 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
