siRNA Details
Gene Symbol: PRR36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190467.1 | NP_001177396.1 | 5 | 2376 | ||
GenBank mRNA | AK025837.1 | - | 2 | 181 | ||
AK096643.1 | - | 2 | 457 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
