siRNA Details
Gene Symbol: RFPL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013734.2 | NP_001013756.2 | 3 | 1389 | ||
XM_017010897.1 | XP_016866386.1 | 4 | 1531 | |||
XM_017010898.1 | XP_016866387.1 | 4 | 1706 | |||
GenBank mRNA | AK122906.1 | - | 3 | 1389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
