siRNA Details
Gene Symbol: SYF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015484.4 | NP_056299.1 | 3, 4 | 309 | ||
GenBank mRNA | AF273089.1 | - | 3, 4 | 269 | ||
AK303582.1 | - | 3, 4 | 289 | |||
AK314859.1 | - | 3, 4 | 289 | |||
AL080166.1 | - | 3, 4 | 270 | |||
BC010862.1 | - | 3, 4 | 280 | 3877165 | ||
BI668848.1 | - | 3, 4 | 307 | |||
DR155437.1 | - | 3, 4 | 311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
