siRNA Details
Gene Symbol: PLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170585.1 | NP_001164056.1 | 50 | 3651 | ||
NM_153021.4 | NP_694566.4 | 51 | 3684 | |||
XM_011532579.2 | XP_011530881.1 | 53 | 3991 | |||
XM_011532581.2 | XP_011530883.1 | 53 | 3954 | |||
XM_011532584.2 | XP_011530886.1 | 53 | 3991 | |||
XM_011532588.2 | XP_011530890.1 | 53 | 3993 | |||
XM_011532589.2 | XP_011530891.1 | 53 | 3988 | |||
XM_011532590.2 | XP_011530892.1 | 53 | 3993 | |||
XM_011532591.2 | XP_011530893.1 | 53 | 3990 | |||
XM_011532593.2 | XP_011530895.1 | 49 | 3566 | |||
XM_011532601.2 | XP_011530903.1 | 39 | 2979 | |||
XM_011532602.2 | XP_011530904.1 | 38 | 2951 | |||
XM_011532603.2 | XP_011530905.1 | 38 | 3028 | |||
XM_011532609.2 | XP_011530911.1 | 36 | 2782 | |||
XM_011532610.2 | XP_011530912.1 | 28 | 2968 | |||
XM_017003432.1 | XP_016858921.1 | 52 | 3960 | |||
XM_017003433.1 | XP_016858922.1 | 52 | 3926 | |||
XM_017003434.1 | XP_016858923.1 | 53 | 3995 | |||
GenBank mRNA | BC042674.1 | - | 11 | 730 | ||
BC150633.1 | - | 51 | 3684 | 9057003 | ||
BC153864.1 | - | 50 | 3603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
