siRNA Details
Gene Symbol: PLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170585.1 | NP_001164056.1 | 56 | 4137 | ||
NM_153021.4 | NP_694566.4 | 57 | 4170 | |||
XM_011532579.2 | XP_011530881.1 | 59 | 4488 | |||
XM_011532581.2 | XP_011530883.1 | 59 | 4440 | |||
XM_011532589.2 | XP_011530891.1 | 59 | 4474 | |||
XM_011532593.2 | XP_011530895.1 | 55 | 4052 | |||
XM_011532601.2 | XP_011530903.1 | 45 | 3465 | |||
XM_011532602.2 | XP_011530904.1 | 44 | 3437 | |||
XM_011532603.2 | XP_011530905.1 | 44 | 3514 | |||
XM_011532609.2 | XP_011530911.1 | 42 | 3268 | |||
XM_011532610.2 | XP_011530912.1 | 34 | 3454 | |||
XM_017003432.1 | XP_016858921.1 | 58 | 4446 | |||
XM_017003433.1 | XP_016858922.1 | 58 | 4412 | |||
GenBank mRNA | BC042674.1 | - | 17 | 1216 | ||
BC150633.1 | - | 57 | 4170 | 9057003 | ||
BC153864.1 | - | 56 | 4089 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
