siRNA Details
Gene Symbol: CCDC159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080503.2 | NP_001073972.2 | 3 | 215 | ||
XM_006722643.1 | XP_006722706.1 | 4 | 266 | |||
XM_011527678.2 | XP_011525980.2 | 3 | 384 | |||
XM_017026255.1 | XP_016881744.1 | 4 | 266 | |||
XM_017026256.1 | XP_016881745.1 | 4 | 505 | |||
XM_017026257.1 | XP_016881746.1 | 4 | 329 | |||
XM_017026258.1 | XP_016881747.1 | 3 | 377 | |||
XM_017026259.1 | XP_016881748.1 | 3 | 375 | |||
XM_017026260.1 | XP_016881749.1 | 3 | 384 | |||
XM_017026261.1 | XP_016881750.1 | 4 | 507 | |||
XM_017026262.1 | XP_016881751.1 | 3 | 462 | |||
XM_017026263.1 | XP_016881752.1 | 3 | 455 | |||
XM_017026264.1 | XP_016881753.1 | 4 | 507 | |||
GenBank mRNA | AK293611.1 | - | 3 | 164 | ||
AK301653.1 | - | 3 | 196 | |||
AK303641.1 | - | 3 | 179 | |||
AK316339.1 | - | 3 | 189 | |||
BC038439.1 | - | 6 | 958 | |||
CA848583.1 | - | 3 | 185 | |||
DC416232.1 | - | 3 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
