siRNA Details
Gene Symbol: AFMID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010982.4 | NP_001010982.2 | 8 | 667 | ||
NM_001145526.2 | NP_001138998.1 | 8 | 667 | |||
XM_011524326.2 | XP_011522628.1 | 7 | 599 | |||
XM_011524327.2 | XP_011522629.1 | 9 | 835 | |||
XM_011524328.2 | XP_011522630.1 | 9 | 807 | |||
XM_011524329.2 | XP_011522631.1 | 9 | 849 | |||
XM_011524330.2 | XP_011522632.1 | 9 | 853 | |||
XM_011524331.2 | XP_011522633.1 | 8 | 749 | |||
XM_011524332.2 | XP_011522634.1 | 8 | 721 | |||
XM_011524333.2 | XP_011522635.1 | 8 | 626 | |||
XM_011524334.2 | XP_011522636.1 | 6 | 578 | |||
XM_011524335.2 | XP_011522637.1 | 6 | 578 | |||
GenBank mRNA | BC132824.1 | - | 8 | 667 | 40146846 | |
BX648442.1 | - | 8 | 623 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
