siRNA Details
Gene Symbol: RGAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024455.3 | NP_001019626.1 | 1 | 2761 | ||
GenBank mRNA | AB082532.1 | - | 1 | 2761 | ||
AL359579.2 | - | 1 | 1805 | |||
BC151224.1 | - | 1 | 2782 | 9007468 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
