siRNA Details
Gene Symbol: KPNA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516215.2 | XP_011514517.1 | 10 | 1729 | ||
XM_017012208.1 | XP_016867697.1 | 10 | 1729 | |||
XM_017012209.1 | XP_016867698.1 | 10 | 1717 | |||
XM_017012210.1 | XP_016867699.1 | 10 | 1805 | |||
XM_017012211.1 | XP_016867700.1 | 10 | 1729 | |||
XM_017012212.1 | XP_016867701.1 | 10 | 1519 | |||
XM_017012213.1 | XP_016867702.1 | 10 | 1857 | |||
XM_017012214.1 | XP_016867703.1 | 10 | 1433 | |||
XM_017012215.1 | XP_016867704.1 | 9 | 1546 | |||
XM_017012216.1 | XP_016867705.1 | 7 | 1177 | |||
GenBank mRNA | EU126604.1 | - | 9 | 1373 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
