siRNA Details
Gene Symbol: IGSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080444.1 | NP_001073913.1 | 9 | 1361 | ||
XM_011529472.1 | XP_011527774.1 | 9 | 2634 | |||
XM_011529473.1 | XP_011527775.1 | 8 | 2612 | |||
GenBank mRNA | AK092516.1 | - | 9 | 1361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
