siRNA Details
Gene Symbol: LOC101927905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120454.1 | 1 | 431 | |||
NR_120455.1 | 1 | 431 | ||||
GenBank mRNA | AL080194.1 | - | 1 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
