siRNA Details
Gene Symbol: MGC27345 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046216.1 | 1 | 3081 | |||
GenBank mRNA | BC024231.1 | - | 1 | 336 | 4670552 |
Gene Symbol: RBM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011516370.2 | XP_011514672.1 | 20 | 8616 | ||
XM_017012389.1 | XP_016867878.1 | 18 | 8392 | |||
XM_017012390.1 | XP_016867879.1 | 17 | 8284 | |||
GenBank mRNA | AK001384.1 | - | 1 | 2051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
