siRNA Details
Gene Symbol: ADAM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001569.3 | 16 | 1962 | |||
NR_024106.2 | 17 | 2057 | ||||
NR_024107.2 | 18 | 2208 | ||||
NR_073423.1 | 18 | 2305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
