siRNA Details
Gene Symbol: PRR32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122716.1 | NP_001116188.1 | 2 | 255 | ||
GenBank mRNA | BG200532.1 | - | 1 | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
