siRNA Details
Gene Symbol: ZNF284 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037813.2 | NP_001032902.1 | 5 | 647 | ||
XM_011526907.2 | XP_011525209.1 | 5 | 688 | |||
XM_011526908.2 | XP_011525210.1 | 6 | 748 | |||
GenBank mRNA | AK308295.1 | - | 11 | 2353 | ||
AY166789.1 | - | 3 | 349 | |||
AY458845.1 | - | 5 | 647 | |||
CR936662.1 | - | 5 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
