siRNA Details
Gene Symbol: TMEM75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103558.1 | 1 | 868 | |||
GenBank mRNA | AK093424.1 | - | 1 | 868 | ||
AX748117.1 | - | 1 | 868 | |||
BC137383.1 | - | 1 | 490 | 9021381 | ||
BC137385.1 | - | 1 | 490 | 9021383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
