siRNA Details
Gene Symbol: PTRHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013663.1 | NP_001013685.1 | 2 | 352 | ||
GenBank mRNA | BC073803.1 | - | 2 | 352 | 4896064 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
