siRNA Details
Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024496.1 | 2 | 343 | |||
NR_135010.1 | 2 | 198 | ||||
GenBank mRNA | BC042539.1 | - | 2 | 343 | ||
BC068605.1 | - | 2 | 197 | |||
BC104192.2 | - | 1 | 478 | |||
BC104193.2 | - | 1 | 478 | |||
BC112986.1 | - | 1 | 478 | |||
BC112987.1 | - | 1 | 478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
