siRNA Details
Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024496.1 | 1, 2 | 306 | |||
GenBank mRNA | BC042539.1 | - | 1, 2 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
