siRNA Details
Gene Symbol: RNASE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012975.1 | NP_001012993.1 | 1 | 125 | ||
GenBank mRNA | AK296766.1 | - | 4 | 613 | ||
AY665805.1 | - | 1 | 125 | |||
BC133002.1 | - | 1 | 144 | 40147024 | ||
BC133028.1 | - | 1 | 144 | 40147050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
