siRNA Details
Gene Symbol: TMEM249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252402.2 | NP_001239331.1 | 4, 5 | 703 | ||
NM_001252404.2 | NP_001239333.1 | 3, 4 | 608 | |||
NM_001280561.1 | NP_001267490.1 | 5, 6 | 1307 | |||
NR_047684.2 | 4, 5 | 705 | ||||
GenBank mRNA | AB196634.1 | - | 5, 6 | 1307 | ||
BC127763.1 | - | 3, 4 | 522 | |||
BC141966.1 | - | 4, 5 | 618 | |||
BC150569.1 | - | 3, 4 | 522 | |||
BC150570.1 | - | 3, 4 | 522 | |||
BX648457.1 | - | 2 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
