siRNA Details
Gene Symbol: TTC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010854.1 | NP_001010854.1 | 16 | 1973 | ||
NM_001320421.1 | NP_001307350.1 | 16 | 1969 | |||
XM_005267367.1 | XP_005267424.1 | 12 | 1284 | |||
XM_011536497.1 | XP_011534799.1 | 16 | 2031 | |||
XM_017021044.1 | XP_016876533.1 | 16 | 1937 | |||
XM_017021045.1 | XP_016876534.1 | 16 | 1937 | |||
XM_017021046.1 | XP_016876535.1 | 16 | 2031 | |||
XM_017021047.1 | XP_016876536.1 | 15 | 1648 | |||
GenBank mRNA | AK126571.1 | - | 7 | 706 | ||
AL832848.1 | - | 15 | 1651 | |||
BC035865.2 | - | 10 | 1074 | |||
BC048270.2 | - | 10 | 1074 | |||
BX247966.1 | - | 2 | 261 | |||
BX248275.1 | - | 12 | 1265 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
