siRNA Details
Gene Symbol: TTC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010854.1 | NP_001010854.1 | 20 | 3218 | ||
NM_001320421.1 | NP_001307350.1 | 21 | 3265 | |||
XM_005267367.1 | XP_005267424.1 | 18 | 2742 | |||
XM_011536497.1 | XP_011534799.1 | 22 | 3489 | |||
XM_017021044.1 | XP_016876533.1 | 22 | 3395 | |||
XM_017021045.1 | XP_016876534.1 | 21 | 3233 | |||
XM_017021046.1 | XP_016876535.1 | 20 | 3276 | |||
XM_017021047.1 | XP_016876536.1 | 19 | 2893 | |||
GenBank mRNA | AK126571.1 | - | 11 | 1951 | ||
AL832848.1 | - | 19 | 2894 | |||
BC035865.2 | - | 15 | 2514 | |||
BX247966.1 | - | 7 | 1557 | |||
BX248275.1 | - | 16 | 2510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
