siRNA Details
Gene Symbol: TTC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010854.1 | NP_001010854.1 | 3, 4 | 565 | ||
NM_001320421.1 | NP_001307350.1 | 3, 4 | 561 | |||
XM_011536497.1 | XP_011534799.1 | 3, 4 | 623 | |||
XM_017021044.1 | XP_016876533.1 | 3, 4 | 529 | |||
XM_017021045.1 | XP_016876534.1 | 3, 4 | 529 | |||
XM_017021046.1 | XP_016876535.1 | 3, 4 | 623 | |||
XM_017021047.1 | XP_016876536.1 | 3, 4 | 362 | |||
GenBank mRNA | AL832848.1 | - | 2, 3 | 243 | ||
BQ426031.1 | - | 480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
