siRNA Details
Gene Symbol: GCOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285900.3 | NP_001272829.1 | 13 | 1745 | ||
NR_104371.3 | 15 | 2535 | ||||
GenBank mRNA | AY353058.1 | - | 15 | 2406 | ||
JF419331.1 | - | 13 | 1635 |
Gene Symbol: POLR2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015532.4 | NP_056347.1 | 2 | 745 | ||
GenBank mRNA | AF326773.1 | - | 2 | 435 | ||
AK074767.1 | - | 2 | 554 | |||
AK074955.1 | - | 2 | 549 | |||
AK223571.1 | - | 2 | 544 | |||
AK308873.1 | - | 2 | 725 | |||
AY339380.1 | - | 2 | 563 | |||
BC001510.2 | - | 2 | 535 | 3504557 | ||
BI833260.1 | - | 2 | 722 | |||
BQ072611.1 | - | 2 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
