siRNA Details
Gene Symbol: MTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167741.1 | NP_001161213.1 | 1, 2 | 115 | ||
XM_017009440.1 | XP_016864929.1 | 1, 2 | 109 | |||
GenBank mRNA | BX538064.1 | - | 1, 2 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
