siRNA Details
Gene Symbol: MTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010891.4 | NP_001010891.4 | 5 | 471 | ||
NM_001167741.1 | NP_001161213.1 | 4 | 401 | |||
XM_005248495.3 | XP_005248552.1 | 5 | 473 | |||
XM_017009440.1 | XP_016864929.1 | 4 | 395 | |||
GenBank mRNA | AK296864.1 | - | 5 | 466 | ||
BC150295.1 | - | 5 | 461 | |||
BX538064.1 | - | 4 | 381 | |||
DB456588.1 | - | 5 | 471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
