siRNA Details
Gene Symbol: BMS1P20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027293.2 | 11 | 1874 | |||
GenBank mRNA | AB063986.1 | - | 1 | 158 | ||
AB363340.1 | - | 1 | 158 | |||
AB363397.1 | - | 1 | 158 | |||
AF103652.1 | - | 1 | 138 | |||
AF103699.1 | - | 1 | 126 | |||
AJ233731.1 | - | 1 | 149 | |||
AJ399840.1 | - | 1 | 158 | |||
AK090629.1 | - | 11 | 1814 | |||
AM181147.1 | - | 1 | 158 | |||
AY043113.1 | - | 1 | 158 | |||
AY043142.1 | - | 1 | 158 | |||
DB472458.1 | - | 2 | 232 | |||
DQ098732.1 | - | 1 | 158 | |||
DQ490727.1 | - | 1 | 158 | |||
DQ840929.1 | - | 1 | 158 | |||
DQ841007.1 | - | 1 | 158 | |||
KJ134805.1 | - | 1 | 158 | |||
KU602548.1 | - | 1 | 158 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
