siRNA Details
Gene Symbol: CCDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030771.1 | NP_110398.1 | 6 | 1078 | ||
GenBank mRNA | AF301222.1 | - | 3 | 321 | ||
AF382034.1 | - | 6 | 965 | |||
BC029421.1 | - | 6 | 751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
