siRNA Details
Gene Symbol: SH2D7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101404.1 | NP_001094874.1 | 5 | 1194 | ||
XM_011521895.2 | XP_011520197.1 | 5 | 2486 | |||
XM_011521896.2 | XP_011520198.1 | 6 | 1170 | |||
XM_011521899.2 | XP_011520201.1 | 4 | 1961 | |||
XM_017022487.1 | XP_016877976.1 | 6 | 1403 | |||
XM_017022488.1 | XP_016877977.1 | 4 | 978 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
